Learn More
SLC13A3 Polyclonal Antibody
GREENER_CHOICE
SLC13A3 Polyclonal Antibody
Brand:  Invitrogen PA5110375
Product Code. 17138303
  • £442.00 / 100µL
Description
SLC13A3 Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC13A3
Polyclonal
Unconjugated
SLC13A3
hNaDC3; hypothetical protein zgc:77173; mNaDC3; Na(+)/dicarboxylate cotransporter 3; Nadc3; NaDC-3; rNaDC3; SDCT2; slc13a3; sodium-dependent high affinity dicarboxylate transporter 3; sodium-dependent high-affinity dicarboxylate transporter 2; sodium-dependent high-affinity dicarboxylate transporter 3; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3; solute carrier family 13 member 3; zgc:77173
Rabbit
Affinity Chromatography
RUO
64849
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.45 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q8WWT9
SLC13A3
Recombinant fusion protein containing a sequence corresponding to amino acids 160-240 of human SLC13A3 (NP_073740.2).