Learn More
Invitrogen™ gp210 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5110051
Product Code. 17137983
  • £440.00 / 100µL
Description
Immunogen sequence: QNVTAEDTDS IGLALFAPHS SRNYQQHWIL VTCQALGEQV IALSVGNKPS LTNPFPAVEP AVVKFVCAPP SRLTLAPVYT SPQLDMSCPL LQQNKQVVPV SSHRNPRLDL AAYDQEGRRF DNFSSLSIQW ESTRPVLASI EPELPMQLVS QDDESGQKKL HGLQAILVHE ASGTTAITAT ATGYQESHLS SARTKQPHDP L.
NUP210 nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
gp210
Polyclonal
Unconjugated
NUP210
9830001L10; AI836801; FLJ22389; gp190; GP210; integral membrane nucleoporin gp210; Kiaa0906; Nuclear envelope pore membrane protein POM 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nuclear pore protein gp210; nucleoporin 210; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; nucleoporin 210kDa L homeolog; nucleoporin Nup210; NUP210; nup210.L; POM210; pore membrane protein of 210 kDa; PSEC0245; XELAEV_18001839mg
Rabbit
Affinity chromatography
RUO
23225, 54563, 58958
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
0.87 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
P11654, Q8TEM1, Q9QY81
NUP210
Recombinant fusion protein containing a sequence corresponding to amino acids 680-880 of human NUP210 (NP_079199.2).