Learn More
Invitrogen™ FKBP8 Recombinant Rabbit Monoclonal Antibody (2J7P6)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA535702
Product Code. 17116363
  • £438.00 / 100µL
Description
Immunogen sequence: MASCAEPSEP SAPLPAGVPP LEDFEVLDGV EDAEGEEEEE EEEEEEDDLS ELPPLEDMGQ PPAEEAEQPG ALAREFLAAM EPEPAPAPAP EEWLDILGNG.
FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FKBP8
Recombinant Monoclonal
0.4 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O35465, Q14318, Q3B7U9
FKBP8
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FKBP38/FKBP8 (Q14318).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot, Immunocytochemistry
2J7P6
Unconjugated
FKBP8
38 kDa FK506-binding protein; 38 kDa FKBP; 38kDa; FK506 binding protein 8; FK506 binding protein 8, 38kDa; FK506-binding protein 38; FK506-binding protein 8; FK506-binding protein 8 (38kD); FKBP prolyl isomerase 8; FKBP38; FKBP-38; FKBP8; FKBP-8; FKBPR38; hFKBP38; mFKBP38; peptidyl-prolyl cis-trans isomerase FKBP8; PPIase FKBP8; rotamase; Sam11
Rabbit
Affinity chromatography
RUO
14232, 23770, 290652
-20°C, Avoid Freeze/Thaw Cycles