Learn More
Invitrogen™ HAI-1 Recombinant Rabbit Monoclonal Antibody (016)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA529818
Product Code. 17109085
  • £389.00 / 100µL
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human HAI-1/SPINT1.
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HAI-1
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
O43278
SPINT1
Recombinant Human HAI-1/SPINT1 Protein (Met1-Val433).
100 μL
Primary
Human
Antibody
IgG
ELISA
016
Unconjugated
SPINT1
HAI; HAI1; HAI-1; hepatocyte growth factor activator inhibitor 1; hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; kunitz-type protease inhibitor 1; MANSC2; serine peptidase inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 1; Spint1; UNQ223/PRO256
Rabbit
Protein A
RUO
6692
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.