Learn More
ZBP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13285-1-AP-150UL
This item is not returnable.
View return policy
Description
Z-DNA binding protein 1 (ZBP1) belongs to a family of proteins that contain the Zalpha domain which binds specifically to left-handed Z-DNA and Z-RNA. ZBP1 was initially identified as a novel gene that was up-regulated in activated macrophages in mice bearing ascites tumors, suggesting that is may play a role in processes such as host response in neoplasia. More recent reports indicate that the cytosolic ZBP1 has can act like the toll-like receptor TLR9 by detecting cytosolic double-stranded (ds) DNA and trigger induction of type I interferon and other innate immune responses. It is thought that the binding of ZBP1 to dsDNA enhances its association with innate immune response proteins such as the IRF3 transcription factor and the serine/threonine kinase TBK1 (also known as NAK). Multiple isoforms of ZBP1 are known to exist.Specifications
| ZBP1 | |
| Polyclonal | |
| Unconjugated | |
| ZBP1 | |
| C20orf183, DAI, DLM 1, DLM1, Z DNA binding protein 1, ZBP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 81030 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H171 | |
| ZBP1 | |
| ZBP1 Fusion Protein Ag4119 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |