Learn More
Alpha E catenin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12831-1-AP-20UL
 View more versions of this product
Product Code. 16870754
  • £135.00 / 20µL
Description
Associates with the cytoplasmic domain of a variety of cadherins. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties. Can associate with both E- and N-cadherins. Originally believed to be a stable component of E-cadherin/catenin adhesion complexes and to mediate the linkage of cadherins to the actin cytoskeleton at adherens junctions. In contrast, cortical actin was found to be much more dynamic than E-cadherin/catenin complexes and CTNNA1 was shown not to bind to F-actin when assembled in the complex suggesting a different linkage between actin and adherence junctions components. The homodimeric form may regulate actin filament assembly and inhibit actin branching by competing with the Arp2/3 complex for binding to actin filaments. May play a crucial role in cell differentiation.
Specifications
Specifications
Alpha E catenin
Polyclonal
Unconjugated
CTNNA1
alpha catenin, Alpha E catenin, Cadherin associated protein, CAP102, Catenin alpha 1, CTNNA1, alpha-catenin, alpha-E-catenin, alphaE-catenin
Rabbit
Antigen Affinity Chromatography
RUO
12385, 1495, 307505
-20°C
Liquid
Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P26231, P35221
Ctnna1
Alpha E catenin Fusion Protein Ag3565
20 μL
Primary
Human, Rat, Mouse