Learn More
ROBU VitraPOR™ Pipeline Filter
Click to view available options
Diameter (Metric):
30 mm
60 mm
Porosity:
0
1
2
3
4
5
Unit Size:
Each
Pack of 2
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Sterility | Non-sterile |
| Includes | Tube Adapters |
| No. per Pack | 2/Pk. |
| Porosity | 0 |
| Length (Metric) | 140 mm |
| Diameter (Metric) | 30 mm |