Learn More
Kallistatin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11430-2-AP-150UL
This item is not returnable.
View return policy
Description
SERPINA4 inhibits human amidolytic and kininogenase activities of tissue kallikrein. The inhibition is achieved by formation of an equimolar, heat- and SDS-stable complex between the inhibitor and the enzyme, and generation of a small C-terminal fragment of the inhibitor due to cleavage at the reactive site by tissue kallikrein.Specifications
| Kallistatin | |
| Polyclonal | |
| Unconjugated | |
| SERPINA4 | |
| KAL, Kallikrein inhibitor, kallistatin, KLST, KST, Peptidase inhibitor 4, PI 4, PI4, Serpin A4, SERPINA4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5267 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P29622 | |
| SERPINA4 | |
| Kallistatin Fusion Protein Ag1962 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |