Learn More
A&D Weighing Ion BM Series Balance
Click to view available options
Capacity:
2.1 g/5.2 g
22 g
220 g
320 g
5.1 g/22 g
5.2 g
520 g
Readability:
0.001 mg
0.001 mg/0.01 mg
0.01 mg
0.1 mg
1 μg
Unit Size:
Each
Description
- Easy, fanless elimination of static electricity using a built-in ionizer
- Anti-static breeze break
- Comes equipped with the AD-1676 tabletop breeze break (medium) as standard (BM-5/5D)
- Large weighing chamber for improved usability
- Built-in temperature, humidity, and atmospheric pressure sensors
- Interlockable slide doors allowing efficient operation with both hands
- Motorized calibration with an internal mass:
- Automatically triggered based on temperature variation
- Triggered by operator
- Clearly visible, white-backlit LCD
- Memory: up to 200 data sets
- AD-1688 as standard accessory: dedicated USB key to record weighing data as well as environmental parameters into MS Excel readable files
- 12 weighing units: g, mg, oz, ozt, ct, mom, dwt, GN, tael, tola, pcs (counting), %
- Special pans/holders for BM-5/5D/20/22: Small pans for sampling minute amounts, holders for PM filters, holders for microfuge tubes
- Optional pipette accuracy testing kit
- Interfaces: RS-232C (standard), Quick USB (standard), Ethernet (optional)
Specifications
Specifications
| Certifications/Compliance | GLP/GMP/GCP/ISO |
| Connection Type | RS-232 C and Quick USB |
| Capacity | 5.2 g |
| Display Type | Reverse-backlit LCD |
| Readability | 1 μg |
| Linearity | ±0.010 mg |
| Model | BM-5 |
| Repeatability | 0.0012 mg (for 1 g) |
| Stabilization Time | 10 seconds |
| Type | Micro Analytical Balance |
| Show More |