Learn More
PINX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12368-1-AP-20UL
 View more versions of this product
Product Code. 16850054
  • £135.00 / 20µL
Description
Valosin containing protein (VCP), also designated TERA (for transitional endoplasmic reticulum ATPase), is a member of the AAA family of ATPases, which are involved in a variety of cellular activities. VCP is involved in a variety of membrane functions and in the regulation of the cell cycle. VCP associates with ubiquitinated IkB-a as well as with the 26S proteosome, indicating a potential role for VCP in the proteosome-mediated degradation of IkB-a. NVL (nuclear valosin-containing protein-like), also known as NVLp, is an 856 amino acid nuclear protein belonging to the AAA ATPase family. Implicated in ATP-dependent nuclear processes and ribosome synthesis, NVL exists as three alternatively spliced isoforms designated NVL isoform 1 (NVLp.2), NVL isoform 2 (NVLp.1) and NVL isoform 3. Widely expressed, NVL is found at highest levels in pancreas, retina, heart, skeletal muscle and placenta.
Specifications
Specifications
PINX1
Polyclonal
Unconjugated
PINX1
LPTL, LPTS, PIN2 interacting protein 1, Pin2 interacting protein X1, PINX1, Protein 67 11 3, TRF1 interacting protein 1
Rabbit
Antigen Affinity Chromatography
RUO
305963, 54984, 72400
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A4L691, Q96BK5, Q9CZX5
PINX1
PINX1 Fusion Protein Ag3043
20 μL