Learn More
LARP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17067-1-AP-150UL
 View more versions of this product
Product Code. 16846894
  • £349.00 / 150µL
Description
LARP7 gene ontology annotations related to this gene include RNA processing.
Specifications
Specifications
LARP7
Polyclonal
Unconjugated
LARP7
HDCMA18P, La related protein 7, LARP7, PIP7S
Rabbit
Antigen Affinity Chromatography
RUO
28036, 51574, 686883
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q05CL8, Q4G0J3, Q5XI01
LARP7
LARP7 Fusion Protein Ag10891
150 μL