Learn More
ACOT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15972-1-AP-150UL
 View more versions of this product
Product Code. 16845394
  • £349.00 / 150µL
Description
This gene encodes a member of the acyl coenzyme family. The encoded protein hydrolyzes the CoA thioester of palmitoyl-CoA and other long-chain fatty acids. Decreased expression of this gene may be associated with mesial temporal lobe epilepsy. Alternatively spliced transcript variants encoding distinct isoforms with different subcellular locations have been characterized.
Specifications
Specifications
ACOT7
Polyclonal
Unconjugated
ACOT7
ACH1, ACOT7, ACT, acyl CoA thioesterase 7, BACH, Brain acyl CoA hydrolase, CTE II, CTE IIa, hBACH, LACH, LACH1
Rabbit
Antigen Affinity Chromatography
RUO
11332, 26759, 70025
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00154, Q64559, Q91V12
Acot7
ACOT7 Fusion Protein Ag8759
150 μL