Learn More
ZNF668 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25669-1-AP-150UL
 View more versions of this product
Product Code. 16834235
  • £349.00 / 150µL
Description
ZNF668 is a new candidate transcription factor.
Specifications
Specifications
ZNF668
Polyclonal
Unconjugated
ZNF668
zinc finger protein 668, ZNF668
Rabbit
Antigen Affinity Chromatography
RUO
79759
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96K58
ZNF668
ZNF668 Fusion Protein Ag22506
150 μL