Learn More
RAB25 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16821284 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16821284 20 μL 20µL
16811284 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16821284 Supplier Proteintech Supplier No. 131891AP20UL
Rabbit Polyclonal Antibody
RAB proteins, such as RAB25, are members of the RAS superfamily of small GTPases that are involved in membrane trafficking. Members of the RAB11 subfamily, including RAB25, control the return of internalized membrane-associated moieties to the cell surface.
Specifications
Antigen RAB25
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAB25
Gene Accession No. P57735
Gene Alias CATX 8, CATX8, RAB25, Ras related protein Rab 25
Gene Symbols RAB25
Host Species Rabbit
Immunogen RAB25 Fusion Protein Ag3864
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57111
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG