Learn More
Texwipe TexBond™ 22 Cleanroom Bond Paper A4
Product Code. 16687863 Shop All Texwipe Products
Click to view available options
Color:
Blue
Pink
White
Yellow
Unit Size:
Each
Pack of 2500
Product Code. 16687863
Brand: Texwipe TX5611
Only null left
Add to basket
Add to basket
Medium-weight cellulose paper saturated with synthetic latex substantially reduces particle generation associated with standard papers.
  • Excellent retention of inks and toners
  • Low particle and ion release
  • Latex-impregnated for optimal purity and smoothness
  • Minimal static charging prevents pages from sticking
  • Sterilisable
  • Weight: 75 g/m²
Specifications
Material Bond Paper
Ruled No
Width (English) Page 8.2 in.
Length (English) Page 11.74 in.
Color Blue
Cover Color Blue