Learn More
Invitrogen™ STC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106379
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Stanniocalcin 2.
Has an anti-hypocalcemic action on calcium and phosphate homeostasis.
Specifications
| STC2 | |
| Polyclonal | |
| Unconjugated | |
| STC2 | |
| AW125853; mustc2; stanniocalcin 2; stanniocalcin-2; Stanniocalcin-related protein; STC2; STC-2; Stc2l; STC-related protein; STCRP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20856, 8614 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O76061, O88452 | |
| STC2 | |
| A synthesized peptide derived from human STC2(Accession O76061), corresponding to amino acid residues P268-R302. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |