Learn More
Invitrogen™ PIBF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102686
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PIBF1.
Mediator of progesterone that by acting on the phospholipase A2 enzyme interferes with arachidonic acid metabolism, induces a Th2 biased immune response, and by controlling NK activity exerts an anti-abortive effect.
Specifications
| PIBF1 | |
| Polyclonal | |
| Unconjugated | |
| PIBF1 | |
| C13orf24; Centrosomal protein of 90 kDa; CEP90; PIBF; PIBF1; progesterone immunomodulatory binding factor 1; progesterone-induced blocking factor 1; progesterone-induced-blocking factor 1; RGD1305077; RP11-505F3.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10464, 306104, 52023 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WXW3 | |
| PIBF1 | |
| A synthesized peptide derived from human PIBF1(Accession Q8WXW3), corresponding to amino acid residues V725-T757. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |