Learn More
Invitrogen™ BTBD6 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA599596
Product Code. 16653845
  • £457.00 / 100µL
Description
Antibody detects endogenous levels of total BTBD6.
Adapter protein for the CUL3 E3 ubiquitin-protein ligase complex. Involved in late neuronal development and muscle formation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BTBD6
Polyclonal
Unconjugated
BTBD6
BDPL; BTB (POZ) domain containing 6; BTB domain containing 6; BTB domain protein BDPL; BTB/POZ domain-containing protein 6; Btbd6; glucocorticoid receptor AF-1 coactivator-1; lens BTB domain protein; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein 6
Rabbit
Affinity Chromatography
RUO
690367, 90135
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q96KE9
BTBD6
A synthesized peptide derived from human BTBD6(Accession Q96KE9), corresponding to amino acid residues G296-Y346.
100 μL
Primary
Human, Rat