Learn More
Invitrogen™ B4GALNT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103653
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total B4GALNT2.
B4GALNT2 catalyzes the last step in the biosynthesis of the human Sd(a) antigen through the addition of an N-acetylgalactosamine residue via a beta-1,4 linkage to a subterminal galactose residue substituted with an alpha-2,3-linked sialic acid. B4GALNT2 also catalyzes the last step in the biosynthesis of the Cad antigen (Montiel et al., 2003 [PubMed 12678917]).
Specifications
| B4GALNT2 | |
| Polyclonal | |
| Unconjugated | |
| B4GALNT2 | |
| AI593864; B4galnt2; B4GALT; beta 1,4 N-acetylgalactosaminyltransferase/betal,4 GalNAcT/Sda-GalNAcT; beta-1,4 N-acetylgalactosaminyltransferase 2; beta-1,4-N-acetyl-galactosaminyl transferase 2; beta-1,4-N-acetyl-galactosaminyltransferase 2; beta-4-N-acetylgalactosaminyltransferase; CT-Gal NAc transferase; Dlb1; Dlb-1; dolichos lectin binding 1; Galg2; Galgt2; Ggm3; Igf2bp1; N-acetylgalactosaminyltransferase; sd(a) beta-1,4-GalNAc transferase; sda beta-1,4-GalNAc transferase; UDP-GalNAc:Neu5Aca2-3Galb-R b1,4-N-acetylgalactosaminyltransferase; UDP-GalNAc:Neu5Acalpha2-3Galbeta-R beta1,4-N-acetylgalactosaminyltransferase; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosyl-N-acetylglucosaminylpolypeptide-beta-1, 4-N-acetylgalactosaminyltransferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 124872, 14422 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q09199, Q8NHY0 | |
| B4GALNT2 | |
| A synthesized peptide derived from human B4GALNT2(Accession Q8NHY0), corresponding to amino acid residues G37-M87. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |