Learn More
Invitrogen™ B3GALT1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101455
Product Code. 16627934
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total B3GALT1.
This gene is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesized by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). This gene is expressed exclusively in the brain. The encoded protein shows strict donor substrate specificity for UDP-galactose. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
B3GALT1
Polyclonal
Unconjugated
B3GALT1
B3galt1; Beta-1,3-galactosyltransferase 1; beta-1,3-GalTase 1; beta3GalT1; beta3Gal-T1; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 1; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase-I; UDP-galactose:beta-N-acetylglucosamine beta1, 3-galactosyltransferase 1; UDP-galactose:beta-N-acetyl-glucosamine-beta-1,3-galactosyltransferase 1
Rabbit
Affinity chromatography
RUO
26877, 8708
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O54904, Q9Y5Z6
B3GALT1
A synthesized peptide derived from human B3GALT1(Accession Q9Y5Z6), corresponding to amino acid residues S6-R56.
100 μL
Primary
Human, Mouse