Learn More
Invitrogen™ SENP7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599191
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SENP7.
SENP7 is a protease that may be involved in two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature conjugatable forms and/or deconjugation of SUMO1, SUMO2 and SUMO3 from targeted substrate proteins.
Specifications
| SENP7 | |
| Polyclonal | |
| Unconjugated | |
| SENP7 | |
| 2410152H17Rik; 2810413I22Rik; 2900036C23Rik; 6030449K19Rik; AI790676; KIAA1707; SENP7; sentrin/SUMO-specific protease SENP7; Sentrin-specific protease 7; SSP2; SUMO1/sentrin specific peptidase 7; SUMO1/sentrin specific protease 7; Sumo1/sentrin/SMT3 specific peptidase 7; SUMO-1-specific protease 2; Susp2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57337, 66315 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BUH8, Q9BQF6 | |
| SENP7 | |
| A synthesized peptide derived from human SENP7(Accession Q9BQF6), corresponding to amino acid residues E962-E1012. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu