Learn More
Invitrogen™ HCN4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104030
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HCN4.
HCN4 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. It has been shown that HCN4 channels function as receptors for sour taste, and are associated with pacemaker potential generation in the sinoatrial node.
Specifications
| HCN4 | |
| Polyclonal | |
| Unconjugated | |
| Hcn4 | |
| Bcng3; BCNG-3; brain cyclic nucleotide-gated channel 3; Hcn3; Hcn4; hyperpolarization activated cyclic nucleotide gated potassium channel 4; hyperpolarization activated cyclic nucleotide-gated cation channel 4; hyperpolarization activated cyclic nucleotide-gated potassium channel 4; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4; SSS2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10021, 330953, 59266 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70507, Q9JKA7, Q9Y3Q4 | |
| Hcn4 | |
| A synthesized peptide derived from human HCN4(Accession Q9Y3Q4), corresponding to amino acid residues G953-P1003. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |