Learn More
Invitrogen™ HLTF Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA597829
Product Code. 16590652
  • £432.00 / 100µg
Description
HLTF Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
This gene encodes a member of the SWI/SNF family. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein contains a RING finger DNA binding motif. Two transcript variants encoding the same protein have been found for this gene. However, use of an alternative translation start site produces an isoform that is truncated at the N-terminus compared to the full-length protein.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HLTF
Polyclonal
Unconjugated
HLTF
AF010600; BC057116; DNA-binding protein/plasminogen activator inhibitor 1 regulator; DNA-binding protein/plasminogen activator inhibitor-1 regulator; helicase like transcription factor; helicase-like transcription factor; HIP116; HIP116A; HLTF; HLTF1; mem; P113; RING finger protein 80; RING-type E3 ubiquitin transferase HLTF; RNF80; Smarca3; Snf2l3; SNF2-like 3; Sucrose nonfermenting protein 2-like 3; sucrose nonfermenting-like 3; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 3; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 3; TNF-response element-binding protein; ZBU1
Rabbit
Protein G
RUO
6596
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
3.1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q14527
HLTF
Recombinant Human Helicase-like transcription factor protein (332-476AA).