Learn More
Invitrogen™ FCF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595887
This item is not returnable.
View return policy
Description
Immunogen sequence: PCITDCVMAE IEKLGQKYRV ALRIAKDPRF ERLPCTHKGT YADDCLVQRV TQHKCYIVAT VDRDLKRRIR KIPGVPIMYI SNHRYNIERM PDDYGAPRF.
Essential protein involved in pre-rRNA processing and 40S ribosomal subunit assembly.
Specifications
| FCF1 | |
| Polyclonal | |
| Unconjugated | |
| FCF1 | |
| Bka; C14orf111; CGI-35; FCF1; FCF1 rRNA-processing protein; FCF1 small subunit; RGD1310899; rRNA-processing protein FCF1 homolog; UTP24 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 299198, 51077 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.87 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9Y324 | |
| FCF1 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-198 of human FCF1 (NP_0570461). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |