Learn More
Capreomycin sulfate, MedChemExpress
Product Code. 16499867
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16499867
Brand: Medchem Express HY17566
MedChemExpress Capreomycin sulfate is a peptide antibiotic, commonly grouped with the aminoglycosides, which is given in combination with other antibiotics for MDR-tuberculosis.
Capreomycin sulfate is a peptide antibiotic, commonly grouped with the aminoglycosides, which is given in combination with other antibiotics for MDR-tuberculosis.
Specifications
Chemical Name or Material Capreomycin sulfate
CAS 1405-37-4
Color White
Purity Grade Notes Research
Molecular Formula C25H46N14O11S
Quantity 5 g
Solubility Information H2O : ≥ 37 mg/mL (49.28 mM)
SMILES O=C(CC(N)CCCN)NCC(C(N/C(C(NC(C1NC(NCC1)=N)C2=O)=O)=C/NC(N)=O)=O)NC(C(NC(C(CN2)N)=O)C)=O.O=S(O)(O)=O
Molecular Weight (g/mol) 750.78
Formula Weight 750.78
Percent Purity 98.07%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation