Learn More
JW 55, MedChemExpress
Product Code. 16492087
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16492087
Brand: Medchem Express HY13968
MedChemExpress JW 55 is a potent and selective β-catenin signaling pathway inhibitor, which functions via inhibition of the PARP domain of tankyrase 1 and tankyrase 2 (TNKS1/2). JW 55 decreases auto-PARsylation of TNKS1/2 in vitro with IC50s of 1.9 μM and 830 nM respectively.
JW 55 is a potent and selective β-catenin signaling pathway inhibitor, which functions via inhibition of the PARP domain of tankyrase 1 and tankyrase 2 (TNKS1/2). JW 55 decreases auto-PARsylation of TNKS1/2 in vitro with IC50s of 1.9 μM and 830 nM respectively.
Specifications
Chemical Name or Material JW 55
CAS 664993-53-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H26N2O5
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (115.08 mM)
SMILES O=C(C1=CC=CO1)NC2=CC=C(C(NCC3(C4=CC=C(OC)C=C4)CCOCC3)=O)C=C2
Molecular Weight (g/mol) 434.48
Formula Weight 434.48
Percent Purity 98.77%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death