Learn More
Decloxizine dihydrochloride, MedChemExpress
Product Code. 16491976
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
Product Code. 16491976
Brand: Medchem Express HYA0075
MedChemExpress Decloxizine dihydrochloride(UCB-1402; NSC289116) is a histamine 1 receptor antagonist.
Decloxizine dihydrochloride(UCB-1402
Specifications
Chemical Name or Material Decloxizine dihydrochloride
Physical Form Solid
CAS 13073-96-6
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C21H30Cl2N2O2
For Use With (Application) COVID-19-immunoregulation
Synonym UCB 1402 dihydrochloride
Solubility Information H2O : ≥ 100 mg/mL (241.91 mM) ∣DMSO : ≥ 50 mg/mL (120.95 mM)
SMILES OCCOCCN1CCN(C(C2=CC=CC=C2)C3=CC=CC=C3)CC1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 413.38
Formula Weight 413.38
Percent Purity 98.77%
Grade Research