Learn More
ML335, MedChemExpress
Product Code. 16490277
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16490277
Brand: Medchem Express HY104005
MedChemExpress ML335 is a selective activator of both TREK-1 and TREK-2.
ML335 is a selective activator of both TREK-1 and TREK-2.
Specifications
Chemical Name or Material ML335
CAS 825658-06-8
Color White
Purity Grade Notes Research
Molecular Formula C15H14Cl2N2O3S
Quantity 50 mg
Solubility Information DMSO : 155 mg/mL (415.27 mM; Need ultrasonic and warming)
SMILES O=C(NCC1=CC=C(Cl)C=C1Cl)C2=CC=C(NS(=O)(C)=O)C=C2
Molecular Weight (g/mol) 373.25
Formula Weight 373.25
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation