Learn More
Gomisin G, MedChemExpress
Product Code. 16489138
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489138 10 mg 10mg
16441548 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16489138 Supplier Medchem Express Supplier No. HYN0858
MedChemExpress Gomisin G is an ethanolic extract of the stems of Kadsura interior; exhibits potent anti-HIV activity with EC50 and therapeutic index (TI) values of 0.006 microgram/mL and 300, respectively.
Gomisin G is an ethanolic extract of the stems of Kadsura interior
Specifications
Chemical Name or Material Gomisin G
CAS 62956-48-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H32O9
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (93.18 mM)
SMILES COC1=C(OCO2)C2=CC([C@H](OC(C3=CC=CC=C3)=O)[C@](O)(C)[C@@H](C)C4)=C1C(C4=CC(OC)=C5OC)=C5OC
Molecular Weight (g/mol) 536.57
Formula Weight 536.57
Percent Purity 99.9%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus