Learn More
JNK-IN-7, MedChemExpress
Product Code. 16486688
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16486688
Brand: Medchem Express HY15617
MedChemExpress JNK-IN-7 is a potent JNK inhibitor with IC50 of 1.5, 2 and 0.7 nM for JNK1, JNK2 and JNK3, respectively.
JNK-IN-7 is a potent JNK inhibitor with IC50 of 1.5, 2 and 0.7 nM for JNK1, JNK2 and JNK3, respectively.
Specifications
Chemical Name or Material JNK-IN-7
Physical Form Solid
CAS 1408064-71-0
Color Gray
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C28H27N7O2
For Use With (Application) Cancer-Kinase/protease
Synonym JNK inhibitor
Solubility Information DMSO : 33.33 mg/mL (67.53 mM; Need ultrasonic)
SMILES O=C(C1=CC(NC(/C=C/CN(C)C)=O)=CC=C1)NC(C=C2)=CC=C2NC3=NC=CC(C4=CN=CC=C4)=N3
Molecular Weight (g/mol) 493.56
Formula Weight 493.56
Percent Purity 98.41%
Grade Research