Learn More
TH588, MedChemExpress
Product Code. 16483326
Encompass_Preferred
Click to view available options
Quantity:
5 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
5mg
Product Code. 16483326
Brand: Medchem Express HY12814
MedChemExpress TH588 is first-in-class nudix hydrolase family inhibitor that potently and selectively engage and inhibit the MTH1 (IC50= 5 nM).
TH588 is first-in-class nudix hydrolase family inhibitor that potently and selectively engage and inhibit the MTH1 (IC50= 5 nM).
Specifications
Chemical Name or Material TH588
CAS 1609960-31-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C13H12Cl2N4
Quantity 25 mg
Solubility Information DMSO : 16 mg/mL (54.21 mM; Need ultrasonic and warming)
SMILES NC1=NC(C2=CC=CC(Cl)=C2Cl)=CC(NC3CC3)=N1
Molecular Weight (g/mol) 295.17
Formula Weight 295.17
Percent Purity 98.56%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death