Learn More
GDC-0927, MedChemExpress
Product Code. 16480777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480777 50 mg 50mg
16493096 25 mg 25mg
16487187 5 mg 5mg
16444468 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16480777 Supplier Medchem Express Supplier No. HY111484
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress GDC-0927 (SRN-927) is a potent, non-steroidal, orally bioavailable, selective estrogen receptor antagonist.
GDC-0927 (SRN-927) is a potent, non-steroidal, orally bioavailable, selective estrogen receptor antagonist.
Specifications
Chemical Name or Material GDC-0927
CAS 1642297-01-5
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C28H28FNO4
Quantity 50 mg
Synonym SRN-927
Solubility Information DMSO : 83.33 mg/mL (180.56 mM; Need ultrasonic)
SMILES OC1=CC=C2C(C(C)=C(C3=CC=CC(O)=C3)[C@H](C4=CC=C(OCCN5CC(CF)C5)C=C4)O2)=C1
Molecular Weight (g/mol) 461.52
Formula Weight 461.52
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death