Learn More
GPR120 modulator 2, MedChemExpress
Product Code. 16480528
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16480528
Brand: Medchem Express HY50172
MedChemExpress GPR120 modulator 2 is a G protein coupled receptor 120 (GPR120) modulator extracted from patent US8394841B2, compound example F13. GPR120 modulator 2 can be used for the research of diseases associated with abnormal or deregulated GPR120, such as diabetes.
GPR120 modulator 2 is a G protein coupled receptor 120 (GPR120) modulator extracted from patent US8394841B2, compound example F13. GPR120 modulator 2 can be used for the research of diseases associated with abnormal or deregulated GPR120, such as diabetes.
Specifications
Chemical Name or Material GPR120 modulator 2
CAS 1050506-87-0
Color White
Purity Grade Notes Research
Molecular Formula C20H18ClNO3S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (257.81 mM)
SMILES O=C(O)CCC1=CC=C(OCC2=NC(C3=CC=CC=C3Cl)=CS2)C=C1C
Molecular Weight (g/mol) 387.88
Formula Weight 387.88
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism