Learn More
BRAF inhibitor, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BRAF inhibitor is a B-Raf inhibitor extracted from patent WO/2011103196 A1, Compound P-0850.
Specifications
Specifications
| Chemical Name or Material | BRAF inhibitor |
| CAS | 918505-61-0 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H18F2N4O3S |
| Quantity | 50 mg |
| Solubility Information | DMSO : 50 mg/mL (109.54 mM; Need ultrasonic) |
| SMILES | CC(C)S(=O)(NC1=CC=C(C(C(C2=CNC3=NC=C(C=C32)C4=CC=CN=C4)=O)=C1F)F)=O |
| Molecular Weight (g/mol) | 456.47 |
| Formula Weight | 456.47 |
| Show More |