Learn More
Trichostatin A, MedChemExpress
Product Code. 16478907
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
Product Code. 16478907
Brand: Medchem Express HY15144
MedChemExpress Trichostatin A (TSA) is a potent and specific inhibitor of HDAC class I/II, with an IC50 value of 1.8 nM for HDAC.
Trichostatin A (TSA) is a potent and specific inhibitor of HDAC class I/II, with an IC50 value of 1.8 nM for HDAC.
Specifications
Chemical Name or Material Trichostatin A
Physical Form Solid
CAS 58880-19-6
Color Light Nude
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H22N2O3
For Use With (Application) Cancer-programmed cell death
Synonym TSA
Solubility Information DMSO : 25 mg/mL (82.68 mM; Need ultrasonic)
SMILES CN(C1=CC=C(C=C1)C([C@@H](/C=C(/C=C/C(NO)=O)C)C)=O)C
Molecular Weight (g/mol) 302.37
Formula Weight 302.37
Percent Purity 95.0%
Grade Research