Learn More
Camostat mesylate, MedChemExpress
Product Code. 16475718
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16475718
Brand: Medchem Express HY1351210mg
MedChemExpress Camostat mesylate (Camostat mesilate) is an orally active, synthetic serine protease inhibitor for chronic pancreatitis. Camostat mesylate, an inhibitor of TMPRSS2, shows antiviral activity against SARS-CoV-2. Camostat mesylate also inhibits the activity of prostasin, trypsin, and matriptase.
Camostat mesylate (Camostat mesilate) is an orally active, synthetic serine protease inhibitor for chronic pancreatitis. Camostat mesylate, an inhibitor of TMPRSS2, shows antiviral activity against SARS-CoV-2. Camostat mesylate also inhibits the activity of prostasin, trypsin, and matriptase.
Specifications
Chemical Name or Material Camostat mesylate
Physical Form Solid
CAS 59721-29-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H26N4O8S
For Use With (Application) Cancer-Kinase/protease
Synonym Camostat mesilate FOY305 FOY-S980
Solubility Information DMSO : 125 mg/mL (252.77 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (101.11 mM)
SMILES O=C(OCC(N(C)C)=O)CC1=CC=C(OC(C2=CC=C(NC(N)=N)C=C2)=O)C=C1.CS(=O)(O)=O
Molecular Weight (g/mol) 494.52
Formula Weight 494.52
Percent Purity 98.45%
Grade Research