Learn More
GW627368, MedChemExpress
Product Code. 16473556
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. 16473556
Brand: Medchem Express HY16963
MedChemExpress GW627368 (GW627368X) is a novel, potent and selective competitive antagonist of prostanoid EP4 receptor with additional human TP receptor affinity, with pKi values of 7.0 and 6.8 for human prostanoid EP4 and TP receptors respectively.
GW627368 (GW627368X) is a novel, potent and selective competitive antagonist of prostanoid EP4 receptor with additional human TP receptor affinity, with pKi values of 7.0 and 6.8 for human prostanoid EP4 and TP receptors respectively.
Specifications
Chemical Name or Material GW627368
CAS 439288-66-1
Color White
Purity Grade Notes Research
Molecular Formula C30H28N2O6S
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (183.61 mM; Need ultrasonic)
SMILES O=C(NS(=O)(C1=CC=CC=C1)=O)CC2=CC=C(N(CC3=C4C(OCC)=C(C=CC=C5)C5=C3OCC)C4=O)C=C2
Molecular Weight (g/mol) 544.62
Formula Weight 544.62
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation