Learn More
Vps34-IN-2, MedChemExpress
Description
Vps34-IN-2 is a novel, potent and selective inhibitor of Vps34 with IC50s of 2 and 82 nM on the Vps34 enzymatic assay and the GFP-FYVE cellular assay, respectively. Vps34-IN-2 shows antiviral activity against SARS-CoV-2 (IC50 of 3.1 μM), HCoV-229E (IC50 of 0.7 μM) and HCoV-OC43.
Specifications
Specifications
| Chemical Name or Material | Vps34-IN-2 |
| Physical Form | Solid |
| CAS | 1523404-29-6 |
| Color | Off-White |
| Quantity | 1 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H25F3N4O3 |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | Ethanol : 50 mg/mL (124.25 mM; Need ultrasonic) |
| SMILES | O=C1C=C(N2[C@H](C)COCC2)N=C3N(CC(C(C)C)=O)[C@H](C(F)(F)F)CCN31 |
| Show More |