Learn More
Etripamil, MedChemExpress
Product Code. 16469897
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16469897
Brand: Medchem Express HY17611
MedChemExpress Etripamil (MSP-2017) is a short-acting L-type calcium-channel antagonist, can be used for the research of Paroxysmal Supraventricular Tachycardia (PSVT). Etripamil (MSP-2017) slows atrioventricular nodal conduction and prolongs atrioventricular nodal refractory periods by inhibiting calcium ion influx through the calcium slow channels in the atrioventricular node cells.
Etripamil (MSP-2017) is a short-acting L-type calcium-channel antagonist, can be used for the research of Paroxysmal Supraventricular Tachycardia (PSVT). Etripamil (MSP-2017) slows atrioventricular nodal conduction and prolongs atrioventricular nodal refractory periods by inhibiting calcium ion influx through the calcium slow channels in the atrioventricular node cells.
Specifications
Chemical Name or Material Etripamil
CAS 1593673-23-4
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C27H36N2O4
Quantity 5 mg
Synonym MSP-2017 (-)-MSP-2017
Solubility Information Ethanol : 120 mg/mL (265.14 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=CC(CCN(CCC[C@](C2=CC=C(OC)C(OC)=C2)(C#N)C(C)C)C)=C1
Molecular Weight (g/mol) 452.59
Formula Weight 452.59
Percent Purity 98.68%
Grade Research
Physical Form Oil
For Use With (Application) COVID-19-immunoregulation