Learn More
CHS-828, MedChemExpress
Product Code. 16469716
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16469716
Brand: Medchem Express HY10079
MedChemExpress CHS-828 (GMX1778) is a competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT), with an IC50 less than 25 nM. CHS-828 (GMX1778) exerts a cytotoxic effect by decreasing the cellular level of NAD+ and exhibits a potent anticancer activity.
CHS-828 (GMX1778) is a competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT), with an IC50 less than 25 nM. CHS-828 (GMX1778) exerts a cytotoxic effect by decreasing the cellular level of NAD+ and exhibits a potent anticancer activity.
Specifications
Chemical Name or Material CHS-828
CAS 200484-11-3
Color White
Purity Grade Notes Research
Molecular Formula C19H22ClN5O
Quantity 50 mg
Synonym GMX1778
Solubility Information DMSO : ≥ 39 mg/mL (104.88 mM)
SMILES N#CN/C(NCCCCCCOC1=CC=C(C=C1)Cl)=N\C2=CC=NC=C2
Molecular Weight (g/mol) 371.86
Formula Weight 371.86
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death