Learn More
Pirodavir, MedChemExpress
Product Code. 16465858
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16465858
Brand: Medchem Express HY13784
MedChemExpress Pirodavir is a potent, broad-spectrum picornavirus inhibitor, and is highly active against both group A and group B rhinovirus serotypes. Pirodavir is very potent in a virus yield reduction assay (IC90=2.3 nM).
Pirodavir is a potent, broad-spectrum picornavirus inhibitor, and is highly active against both group A and group B rhinovirus serotypes.
Specifications
Chemical Name or Material Pirodavir
CAS 124436-59-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H27N3O3
Quantity 10 mg
Synonym R77975
Solubility Information DMSO : 10 mg/mL (27.07 mM; Need ultrasonic)
SMILES O=C(OCC)C1=CC=C(OCCC2CCN(C3=NN=C(C)C=C3)CC2)C=C1
Molecular Weight (g/mol) 369.46
Formula Weight 369.46
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus