Learn More
GSK9311, MedChemExpress
Product Code. 16463138
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463138 10 mg 10mg
16494426 100 mg 100mg
16462636 25 mg 25mg
16409696 50 mg 50mg
16405877 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16463138 Supplier Medchem Express Supplier No. HY100729
MedChemExpress GSK9311, a less active analogue of GSK6853, can be used as a negative control. GSK9311 inhibits BRPF bromodomain with pIC50 values of 6.0 and 4.3 for BRPF1 and BRPF2, respectively.
GSK9311, a less active analogue of GSK6853, can be used as a negative control. GSK9311 inhibits BRPF bromodomain with pIC50 values of 6.0 and 4.3 for BRPF1 and BRPF2, respectively.
Specifications
Chemical Name or Material GSK9311
CAS 1923851-49-3
Color White
Purity Grade Notes Research
Molecular Formula C24H31N5O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (228.56 mM; Need ultrasonic)
SMILES C[C@@H]1CNCCN1C2=C(N(CC)C(C3=CC=CC=C3OC)=O)C=C(C(N4C)=C2)N(C)C4=O
Molecular Weight (g/mol) 437.53
Formula Weight 437.53
Percent Purity 99.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death