Learn More
Kevetrin hydrochloride, MedChemExpress
Product Code. 16463067
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16463067
Brand: Medchem Express HY16271
MedChemExpress Kevetrin hydrochloride is a small molecule and activator of the tumor suppressor protein p53, with potential antineoplastic activity.
Kevetrin hydrochloride is a small molecule and activator of the tumor suppressor protein p53, with potential antineoplastic activity.
Specifications
Chemical Name or Material Kevetrin hydrochloride
Physical Form Solid
CAS 66592-89-0
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C5H10ClN3S
For Use With (Application) Cancer-programmed cell death
Synonym 4-Isothioureidobutyronitrile hydrochloride thioureidobutyronitrile hydrochloride thioureido butyronitrile hydrochloride
Solubility Information H2O : 100 mg/mL (556.58 mM; Need ultrasonic) ∣DMSO : ≥ 40 mg/mL (222.63 mM)
SMILES NC(SCCCC#N)=N.[H]Cl
Molecular Weight (g/mol) 179.67
Formula Weight 179.67
Percent Purity 98.0%
Grade Research