Learn More
Aripiprazole, MedChemExpress
Product Code. 16461419
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
Product Code. 16461419
Brand: Medchem Express HY14546
MedChemExpress Aripiprazole (OPC-14597), an atypical antipsychotic, is a potent and high-affinity dopamine D2 receptor partial agonist. Aripiprazole is an inverse agonist at 5-HT2B and 5-HT2A receptors and displays partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors. Aripiprazole can be used for the research of schizophrenia and COVID19.
Aripiprazole (OPC-14597), an atypical antipsychotic, is a potent and high-affinity dopamine D2 receptor partial agonist. Aripiprazole is an inverse agonist at 5-HT2B and 5-HT2A receptors and displays partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors. Aripiprazole can be used for the research of schizophrenia and COVID19.
Specifications
Chemical Name or Material Aripiprazole
CAS 129722-12-9
Color White
Purity Grade Notes Research
Molecular Formula C23H27Cl2N3O2
Quantity 1 g
Synonym OPC-14597
Solubility Information DMSO : 100 mg/mL (223.02 mM; Need ultrasonic) ∣DMF : 50 mg/mL (111.51 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=CC(OCCCCN3CCN(C4=CC=CC(Cl)=C4Cl)CC3)=C2)CC1
Molecular Weight (g/mol) 448.39
Formula Weight 448.39
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation