Learn More
NH125, MedChemExpress
Product Code. 16459646
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16459646
Brand: Medchem Express HY100576
MedChemExpress NH125 is a potent and selective inhibitor of eukaryotic elongation factor 2 kinase (eEF-2K/CaMKIII), also can induce eEF2 phosphorylation, with an IC50 of 60 nM for eEF-2K.
NH125 is a potent and selective inhibitor of eukaryotic elongation factor 2 kinase (eEF-2K/CaMKIII), also can induce eEF2 phosphorylation, with an IC50 of 60 nM for eEF-2K.
Specifications
Chemical Name or Material NH125
CAS 278603-08-0
Color White
Purity Grade Notes Research
Molecular Formula C27H45IN2
Quantity 50 mg
Solubility Information DMSO : 160 mg/mL (305.02 mM; Need ultrasonic)
SMILES CC1=[N+](CC2=CC=CC=C2)C=CN1CCCCCCCCCCCCCCCC.[I-]
Molecular Weight (g/mol) 524.56
Formula Weight 524.56
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease