Learn More
Ketoconazole, MedChemExpress
Product Code. 16458096
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
Product Code. 16458096
Brand: Medchem Express HYB0105
MedChemExpress Ketoconazole (R-41400) is an imidazole anti-fungal agent, a CYP3A4 and CYP24A1 inhibitor.
Ketoconazole (R-41400) is an imidazole anti-fungal agent, a CYP3A4 and CYP24A1 inhibitor.
Specifications
Chemical Name or Material Ketoconazole
Physical Form Solid
CAS 65277-42-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C26H28Cl2N4O4
For Use With (Application) COVID-19-immunoregulation
Synonym Ketoconazol R 41400
Solubility Information DMSO : 25 mg/mL (47.04 mM; Need ultrasonic)
SMILES CC(N1CCN(C2=CC=C(OC[C@@H]3O[C@@](CN4C=CN=C4)(C5=CC=C(Cl)C=C5Cl)OC3)C=C2)CC1)=O
Molecular Weight (g/mol) 531.43
Formula Weight 531.43
Percent Purity 99.47%
Grade Research