Learn More
RIP2 kinase inhibitor 2, MedChemExpress
Product Code. 16457706
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16457706
Brand: Medchem Express HY19761
MedChemExpress RIP2 kinase inhibitor 2 is a receptor interacting protein-2 (RIP2) kinase inhibitor extracted from patent WO/2014043437 A1, compound example 9.
RIP2 kinase inhibitor 2 is a receptor interacting protein-2 (RIP2) kinase inhibitor extracted from patent WO/2014043437 A1, compound example 9.
Specifications
Chemical Name or Material RIP2 kinase inhibitor 2
CAS 1581270-11-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H28N4O4S
Quantity 100 mg
Solubility Information DMSO : 5 mg/mL (11.56 mM; Need ultrasonic)
SMILES CC(C)(C)S(C1=C(OCCOC)C=C(N=CC=C2NC3=NNC(C)=C3C)C2=C1)(=O)=O
Molecular Weight (g/mol) 432.54
Formula Weight 432.54
Percent Purity 99.16%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease