Learn More
ISRIB (trans-isomer), MedChemExpress
Product Code. 16455896
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16455896
Brand: Medchem Express HY12495
MedChemExpress ISRIB (trans-isomer) is a potent inhibitor of PERK with an IC50 of 5 nM. ISRIB potently reverses the effects of eIF2α phosphorylation (IC50=5 nM).
ISRIB (trans-isomer) is a potent inhibitor of PERK with an IC50 of 5 nM. ISRIB potently reverses the effects of eIF2α phosphorylation (IC50=5 nM).
Specifications
Chemical Name or Material ISRIB (trans-isomer)
CAS 1597403-47-8
Color White
Purity Grade Notes Research
Molecular Formula C22H24Cl2N2O4
Quantity 100 mg
Solubility Information DMSO : 4.55 mg/mL (10.08 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N[C@H]1CC[C@H](NC(COC2=CC=C(Cl)C=C2)=O)CC1)COC3=CC=C(Cl)C=C3.[Relative stereochemistry]
Molecular Weight (g/mol) 451.34
Formula Weight 451.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration