Learn More
Gln-AMS TFA, MedChemExpress
Product Code. 16445546
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16445546
Brand: Medchem Express HY112861A
MedChemExpress Gln-AMS (TFA) is a type Ia aminoacyl-tRNA synthetase (AARS) inhibitor. Gln-AMS inhibits glutaminyl-tRNA synthetase (GlnRS) with a Ki of 1.32 μM.
Gln-AMS (TFA) is a type Ia aminoacyl-tRNA synthetase (AARS) inhibitor. Gln-AMS inhibits glutaminyl-tRNA synthetase (GlnRS) with a Ki of 1.32 μM.
Specifications
Chemical Name or Material Gln-AMS TFA
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H23F3N8O10S
Quantity 100 mg
Solubility Information H2O : 130 mg/mL (220.91 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (84.97 mM; ultrasonic and warming and heat to 60°C)
SMILES O[C@@H]([C@H]([C@H](N1C=NC2=C1N=CN=C2N)O3)O)[C@H]3COS(=O)(NC([C@@H](N)CCC(N)=O)=O)=O.OC(C(F)(F)F)=O
Molecular Weight (g/mol) 588.47
Formula Weight 588.47
Percent Purity 98.73%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation