Learn More
Safinamide, MedChemExpress
Product Code. 16444087
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16444087
Brand: Medchem Express HY70057
MedChemExpress Safinamide is a potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor (IC50=0.098 μM) over MAO-A (IC50=580 μM).
Safinamide also blocks sodium channels and modulates glutamate (Glu) release, showing a greater affinity at depolarized (IC50=8 μM) than at resting (IC50=262 μM) potentials. Safinamide has neuroprotective and neurorescuing effects and can be used for the study of parkinson disease, ischemia stroke etc.al.
Specifications
Chemical Name or Material Safinamide
CAS 133865-89-1
Color White
Purity Grade Notes Research
Molecular Formula C17H19FN2O2
Quantity 50 mg
Synonym FCE 26743 EMD 1195686
Solubility Information DMSO : ≥ 100 mg/mL (330.75 mM)
SMILES O=C([C@@H](NCC1=CC=C(OCC2=CC(F)=CC=C2)C=C1)C)N
Molecular Weight (g/mol) 302.34
Formula Weight 302.34
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation